Learn More
Drostanolone Enanthate, TRC
Specifications
Specifications
| Chemical Name or Material | Drostanolone enanthate |
| CAS | 13425-31-5 |
| Molecular Formula | C27H44O3 |
| Quantity | 250 mg |
| Synonym | Androstan-3-one, 2-methyl-17-[(1-oxoheptyl)oxy]-, (2α,5α,17β)- (9CI), 5α-Androstan-3-one, 17β-hydroxy-2α-methyl-, heptanoate (8CI), Heptanoic acid, ester with 17β-hydroxy-2α-methyl-5α-androstan-3-one, 2α-Methyldihydrotestosterone enanthate |
| InChI Formula | InChI=1S/C27H44O3/c1-5-6-7-8-9-25(29)30-24-13-12-21-20-11-10-19-16-23(28)18(2)17-27(19,4)22(20)14-15-26(21,24)3/h18-22,24H,5-17H2,1-4H3/t18-,19+,20+,21+,22+,24+,26+,27+/m1/s1 |
| SMILES | CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)[C@H](C)C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(2R,5S,8R,9S,10S,13S,14S,17S)-2,10,13-trimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Molecular Weight (g/mol) | 416.6 |
| Formula Weight | 416.6365 |