missing translation for 'onlineSavingsMsg'
Learn More

Dronedarone, MedChemExpress

Product Code. p-200096254
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16418268

Brand: Medchem Express HYA0016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dronedarone (SR 33589), a derivative of amiodarone (HY-14187), is a class III antiarrhythmic agent for the study of atrial fibrillation (AF) and atrial flutter.

Dronedarone is a potent blocker of multiple ion currents, including potassium current, sodium current, and L-type calcium current, and exhibits antiadrenergic effects by noncompetitive binding to β-adrenergic receptors. Dronedarone is a substrate for and a moderate inhibitor of CYP3A4.

Specifications

Chemical Name or Material Dronedarone
CAS 141626-36-0
Color White
Purity Grade Notes Research
Molecular Formula C31H44N2O5S
Quantity 10 mg
Synonym SR 33589
Solubility Information DMSO : ≥ 100 mg/mL (179.61 mM)
SMILES CCCCC1=C(C2=C(C=CC(NS(C)(=O)=O)=C2)O1)C(C3=CC=C(C=C3)OCCCN(CCCC)CCCC)=O
Molecular Weight (g/mol) 556.76
Formula Weight 556.76
Percent Purity 98.43%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.