missing translation for 'onlineSavingsMsg'
Learn More

Dronedarone Hydrochloride, MedChemExpress

Product Code. p-200101063
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498198

Brand: Medchem Express HY75839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+ and Ca2+ currents.

Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+ and Ca2+ currents.

Specifications

Chemical Name or Material Dronedarone Hydrochloride
CAS 141625-93-6
Color White
Purity Grade Notes Research
Molecular Formula C31H45ClN2O5S
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (84.29 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CS(=O)(NC1=CC=C(OC(CCCC)=C2C(C3=CC=C(OCCCN(CCCC)CCCC)C=C3)=O)C2=C1)=O.[H]Cl
Molecular Weight (g/mol) 593.22
Formula Weight 593.22
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.