Learn More
Invitrogen™ DRAQ7™ Dye
Product Code. p-7181019
Click to view available options
Quantity:
200 Assays
50 Assays
Unit Size:
Pack of 200
Pack of 50
Product Code. 16327895
Brand: Invitrogen™ D15106
Edge
DRAQ7 Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry.
This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.
• Stable at room temperature
• Rapid staining of dead cells without wash steps
• No UV excitation and no emission spectral overlap with PE
DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Product Type Dye
Quantity 200 Assays