Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump
Click to view available options
Type:
X-act 5000 Automatic Tube Pump
Includes:
Tube pump, shoulder strap
Unit Size:
Each
Description
- Simple operation and reduced measurement time
- Swiping bar-code loads measurement data
- Automatic adjustment of flow for technical gases
- No external flow meter needed
- Connection with tube swivel
- With segment and matrix display, barcode reader and flashing lights
X-act 5000 Automatic Tube Pump Kit:
- Includes pump with shoulder strap and rechargeable NiMH battery
- With power supply
Specifications
Specifications
| For Use With (Equipment) | Draeger Short-term Tubes and for Sampling Tubes and Systems |
| Type | X-act 5000 Automatic Tube Pump |
| For Use With (Application) | For sampling of toxic gases and vapors in workplace |
| Includes | Tube pump, shoulder strap |