missing translation for 'onlineSavingsMsg'
Learn More

DPH, MedChemExpress

Product Code. p-200099285
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16471217

Brand: Medchem Express HY12070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DPH is a potent cell permeable c-Abl activator, which displays potent enzymatic and cellular activity in stimulating c-Abl activation.

DPH is a potent cell permeable c-Abl activator, which displays potent enzymatic and cellular activity in stimulating c-Abl activation.

Specifications

Chemical Name or Material DPH
CAS 484049-04-9
Color White
Purity Grade Notes Research
Molecular Formula C18H13FN4O2
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (297.34 mM; Need ultrasonic)
SMILES O=C1NC(C(N1)C2=CN(N=C2C3=CC=C(C=C3)F)C4=CC=CC=C4)=O
Molecular Weight (g/mol) 336.32
Formula Weight 336.32
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.