Learn More
Gibco™ DPBS (10X), no calcium, no magnesium
GREENER_CHOICE
Product Code. 11540486
Click to view available options
Quantity:
100mL
500mL
10 x 500 mL
20 x 100 mL
Förpackningsstorlek
100mL
2000mL
5000mL
500mL
Denne vare kan ikke returneres. Se returpolitik
Produktkode 11540486
missing translation for 'mfr': Gibco™ 14200083
Denne vare kan ikke returneres. Se returpolitik
DPBS (10X), no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY
Tekniske data
Chemical Name or Material DPBS
Concentration 10 X
For Use With (Application) Mammalian Cell Culture
pH 6 to 7
Physical Form Liquid
Quantity 10 x 500 mL
Includes No Sodium Pyruvate
Osmolality 2500 to 2900 mOsm/kg
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.