missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DP1 Rabbit anti-Human, Rat, Clone: 8C0O3, Novus Biologicals™
Rabbit Monoclonal Antibody
190.00 EUR - 496.00 EUR
Specifications
| Antigen | DP1 |
|---|---|
| Clone | 8C0O3 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18341094
|
Novus Biologicals
NBP3-16590-20UL |
20 μg |
190.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331256
|
Novus Biologicals
NBP3-16590-100UL |
100 μg |
496.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DP1 Monoclonal antibody specifically detects DP1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| DP1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 311-410 of human DP1 (Q14186). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8C0O3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7027 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title