missing translation for 'onlineSavingsMsg'
Learn More

Doxylamine succinate, 99.46%, MP Biomedicals™

Product Code. 11436822 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 11436822

Brand: MP Biomedicals 0215101710

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 562-10-7
Molecular Formula C21H28N2O5
Molecular Weight (g/mol) 388.46
MDL Number MFCD00056168
InChI Key KBAUFVUYFNWQFM-UHFFFAOYNA-N
Synonym doxylamine succinate, decapryn, evigoa d, a-h injection, decapryn succinate, unisom, alsadorm, gittalun, mereprine
PubChem CID 11224
ChEBI CHEBI:82461
IUPAC Name butanedioic acid; dimethyl({2-[1-phenyl-1-(pyridin-2-yl)ethoxy]ethyl})amine
SMILES OC(=O)CCC(O)=O.CN(C)CCOC(C)(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

Color White to Yellow
Quantity 10 g
Formula Weight 388.5
Percent Purity 0.9946
Physical Form Crystals
Chemical Name or Material Doxylamine succinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.