Learn More
Usbeck™ Double Spoon
Click to view available options
Length (Metric):
120 mm
180 mm
250 mm
300 mm
Unit Size:
Each
Description
- Stainless steel
- Rigid
- Round handle
Specifications
Specifications
| Product Type | Double Spoon End Spatula |
| Width (Metric) Blade | 17/25 mm |
| Material | Stainless Steel |
| Length (Metric) | 120 mm |