Learn More
Thermo Scientific™ Double Layer Tray for Digital Platform Rocker
Shop All Thermo Scientific Products
Click to view available options
Dimensions (L x W):
338 x 299 x 19 mm
Unit Size:
Each
Description
Double platform accommodates more samples while providing consistent rocking motion and easy angle adjustments.
Specifications
Specifications
| Material | Metal |
| Dimensions (L x W) | 338 x 299 x 19 mm |
| Dimensions (L x W x H) | 338 x 299 x 19 mm |
| For Use With (Application) | 88882148, 88882102, 88882103, Various size of tubes, flask, bottle rocking |
| For Use With (Equipment) | Digital Platform Rocker 120V (8888201) or Digital Platform Rocker 230V (88882014) |