missing translation for 'onlineSavingsMsg'
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™

Donkey Polyclonal Antibody has been used in 1 publication

137.00 EUR

Specifications

Antigen IgG (H+L)
Concentration 4.5 mg/mL
Applications Immunoassay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18516182
View Documents
Novus Biologicals
NBP1-75433-0.5mg
0.5 mg
137.00 EUR
0.50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications

Specifications

IgG (H+L)
Immunoassay
Unconjugated
RUO
Sheep
IgG
Affinity purified
Based on IEP, this antibody reacts with heavy gamma chains on sheep IgG and light chains on all sheep immunoglobulins. This antibody has been pre-adsorbed against human and rabbit IgG.
4.5 mg/mL
Polyclonal
Donkey
Adaptive Immunity, Immunology
Purified Sheep IgG (H&L)
Secondary
Store at 4°C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.