missing translation for 'onlineSavingsMsg'
Learn More

DOK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16802604 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16802604 150 μL 150µL
16812604 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16802604 Supplier Proteintech Supplier No. 141081AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DOK-6 (docking protein 6), is a novel member of the Dok-4/5 subclass of the p62 Dok family of intracellular adaptor molecules. Dok-6 is highly expressed in the developing central nervous system and is co-expressed with Ret in several locations, including sympathetic, sensory, and parasympathetic ganglia, as well as in the ureteric buds of the developing kidneys. Pull-down assays using the Dok-6 phosphotyrosine binding (PTB) domain and GDNF-activated Ret indicate that Dok-6 binds to the phosphorylated Ret Tyr(1062) residue. Moreover, ligand activation of Ret resulted in phosphorylation of tyrosine residue(s) located within the unique C terminus of Dok-6 predomitly through a Src-dependent mechanism, indicating that Dok-6 is a substrate of the Ret-Src signaling pathway. (Crowder RJ, Enomoto H, Yang M, Johnson EM Jr, Milbrandt J. Dok-6, a Novel p62 Dok family member, promotes Ret-mediated neurite outgrowth. J Biol Chem. 2004 Oct 1;279(40):42072-81. Epub 2004 Jul 30. It is also known as DOK6, HsT3226, MGC20785 or IRS. 0.

Specifications

Antigen DOK6
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DOK6
Gene Accession No. Q2MHE5, Q6PKX4
Gene Alias docking protein 6, DOK5L, DOK6, HsT3226
Gene Symbols Dok6
Host Species Rabbit
Immunogen DOK6 Fusion Protein Ag5254
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220164, 498898, 623279
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.