Learn More
Docetaxal, TRC
Specifications
Specifications
| Chemical Name or Material | 10-Acetyldocetaxel |
| CAS | 125354-16-7 |
| Molecular Formula | C45 H55 N O15 |
| Quantity | 25 mg |
| Synonym | Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-[[(1,1-dimethylethoxy)carbonyl]amino]-α-hydroxy-, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 10-Acetyldocetaxel, 10-Acetyltaxotere, Docetaxal, PNU 101383, 5β,20-Epoxy-1,7β-dihydroxy-9-oxotax-11-ene-2α,4,10β,13α-tetrayl 4,10-diacetate 2 benzoate 13-[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-3-phenylpropanoate] (10-Acetyldocetaxel) |
| InChI Formula | InChI=1S/C45H55NO15/c1-23-28(58-39(53)33(50)32(26-16-12-10-13-17-26)46-40(54)61-41(4,5)6)21-45(55)37(59-38(52)27-18-14-11-15-19-27)35-43(9,29(49)20-30-44(35,22-56-30)60-25(3)48)36(51)34(57-24(2)47)31(23)42(45,7)8/h10-19,28-30,32-35,37,49-50,55H,20-22H2,1-9H3,(H,46,54)/t28-,29-,30+,32-,33+,34+,35-,37-,43+,44-,45+/m0/s1 |
| SMILES | CC(=O)O[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C1C5(C)C)C |
| Molecular Weight (g/mol) | 849.92 |
| Formula Weight | 849.3572 |