missing translation for 'onlineSavingsMsg'
Learn More

DO3A-Thiol TFA Salt, TRC

Product Code. 30010324
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010324 5 mg 5mg
30008299 1 mg 1mg
30007992 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010324 Supplier Toronto Research Chemicals Supplier No. TRCD4995205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

InChI Formula InChI=1S/C18H33N5O7S.C2HF3O2/c24-15(19-1-10-31)11-20-2-4-21(12-16(25)26)6-8-23(14-18(29)30)9-7-22(5-3-20)13-17(27)28;3-2(4,5)1(6)7/h31H,1-14H2,(H,19,24)(H,25,26)(H,27,28)(H,29,30);(H,6,7)
IUPAC Name 2-[4,7-bis(carboxymethyl)-10-[2-oxo-2-(2-sulfanylethylamino)ethyl]-1,4,7,10-tetrazacyclododec-1-yl]acetic acid;2,2,2-trifluoroacetic acid
Molecular Formula C18 H33 N5 O7 S . x(C2 H F3 O2)
Molecular Weight (g/mol) 577.6
SMILES OC(=O)CN1CCN(CC(=O)O)CCN(CC(=O)NCCS)CCN(CC(=O)O)CC1.OC(=O)C(F)(F)F
Synonym 10-[2-[(2-Mercaptoethyl)amino]-2-oxoethyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic Acid TFA salt, DO3A-Thiol Trifluoroacetate Salt
Chemical Name or Material DO3A-Thiol TFA Salt

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.