Learn More
DO34 analog, MedChemExpress
Product Code. 16474888
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16474888
Brand: Medchem Express HY117771A
MedChemExpress DO34 analog is a triazole DAGL(α) inhibitor extracted from patent WO2017096315 A1, compound 100.
DO34 analog is a triazole DAGL(α) inhibitor extracted from patent WO2017096315 A1, compound 100.
Specifications
Chemical Name or Material DO34 analog
CAS 2098969-71-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H28F3N5O4
Quantity 10 mg
Solubility Information DMSO : ≥ 105 mg/mL (197.54 mM)
SMILES O=C(N1C(CC2=CC=CC=C2)CN(C(OC(C)(C)C)=O)CC1)N3N=CC(C4=CC=C(OC(F)(F)F)C=C4)=N3
Molecular Weight (g/mol) 531.53
Formula Weight 531.53
Percent Purity 96.94%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation