Learn More
STEMCELL Technologies DNase I Solution (1 mg/mL)
Product Code. 15171507
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
15171507 1 mL 1mL
1 options
Product Code. 15171507
Brand: STEMCELL Technologies 07900
Cell dissociation reagent
  • Deoxyribonuclease (DNase) I Solution (1 mg/mL) is useful to reduce or prevent the clumping of concentrated and/or cryopreserved cell suspensions following thawing
Specifications
For Use With (Application) Cell Lysis
Quantity 1 mL