missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB1 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502348S
This item is not returnable.
View return policy
Description
DNAJB1 Monoclonal antibody specifically detects DNAJB1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| DNAJB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BY42 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51507 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| RTF2 | |
| 0610007I11Rik; DjB1; DnaJ (Hsp40) homolog, subfamily B, member 1; DnaJ heat shock protein family (Hsp40) member B1; dnaJ homolog subfamily B member 1; DnaJ protein homolog 1; DNAJ1; Dnajb1; HDJ1; hDj-1; Heat shock 40 kDa protein 1; Heat shock protein; HSP; HSP 40; HSP40; HSPF1; human DnaJ protein 1; NDAJB1; radial spoke 16 homolog B; RSPH16B; Sis1 | |
| Full length human recombinant protein of human C20orf43 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction