Learn More
Melford DNA Markers (Low Range) 250 bp Ladder
Product Code. 30018229
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30018229
Brand: Melford D221001.0
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
  • Store at: 2-8°C (Short Term)
  • Store at: -20°C (Long Term)
Specifications
Quantity 1 mL