missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford DNA Markers (High Range) 1 kb Ladder
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Soluble in: Water
- Store at: 2-8°C
Specifications
Specifications
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction