Learn More
Invitrogen™ DNA Damage Competitive ELISA Kit
Description
The DNA Damage ELISA quantitates PGFM in serum, plasma, saliva, urine, digested DNA, fecal extracts, or cell culture medium. The assay will exclusively recognize both natural and recombinant DNA Damage. Principle of the method The PGFM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Assay Range | 62.6 to 8000 pg/mL |
| Assay Sensitivity | 50.9 pg/mL |
| Description | DNA Damage Competitive ELISA Kit |
| Assay | Multiplex assay on the Luminex Platform |
| Conjugate | HRP |
| Detection Method | Fluorescence |
| Product Type | ELISA |
| Protein Family | Nucleotide |
| Sample Type | Digested DNA, Fecal Extract, Plasma, Saliva, Serum, Supernatant, Urine |
| Sufficient For | 96 assays |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.