Learn More
R&DSystems™ DMEM, high glucose, L-glutamine, sodium pyruvate, no phenol red
Product Code. 18482550
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 18482550
Brand: R&DSystems™ M18650
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dulbecco’s Modified Eagle Medium (DMEM) with 4500 mg/L D-glucose, L-glutamine, and sodium pyruvate. Without phenol red.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell Culture
Quantity 500 mL
Purity or Quality Grade 0.2 μm filtered
Content And Storage 2 - 8 °C, protected from light