missing translation for 'onlineSavingsMsg'
Learn More

Dl-Dithiothreitol, MP Biomedicals™

Product Code. 11436531 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 11436531

Brand: MP Biomedicals 02194821.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to stabilize sulfhydryl-containing enzymes. Dl-Dithiothreitol, MP Biomedicals reduces protein disulfide bonds in sample buffers prior to SDS-PAGE. DTT is also used to reduce the disulfide bridge of the cross-linker N,N'-bis(acryloyl)cystamine to brea

DTT is a Clelands reagent that acts as a protective agent for sulfhydryl groups (-SH). It quantitatively reduces disulfides (-S-S- to -SH). In this reaction the DTT is oxidized to the cyclic disulfide, which ensures the reduction of other disulfides in solution. Disulfide reduction occurs quickly at pH 8.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27565-41-9
Melting Point 40°C to 50°C
pH 4.0 to 6.0 (0.1M in H2O, 25°C) (literature)
Molecular Formula C4H10O2S2
MDL Number MFCD00004877,MFCD00064305
Quantity 250 mg
Synonym 1,4-Dithiothreitol, DTT, 1,4-Dithio-DL-threitol, Cleland Reagent racemic, (±)-threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent
Solubility Information Soluble in water (50mg/mL, clear and colorless solution), ethanol, acetone, ethylate, chloroform and ether.
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
SMILES OC(CS)C(O)CS
IUPAC Name 1,4-disulfanylbutane-2,3-diol
Molecular Weight (g/mol) 154.24
Formula Weight 154.2
Grade Molecular Biology Reagent
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.