missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
Specifications
Specifications
| Color | White |
| Assay Percent Range | ≥99.0% |
| Quantity | 1 g |
| Formula Weight | 154.25 |
| Physical Form | Powder |
| Chemical Name or Material | DL-Dithiothreitol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction