Learn More
DL-Citramalic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | DL-Citramalic Acid |
| CAS | 597-44-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5H8O5 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C5H8O5/c1-5(10,4(8)9)2-3(6)7/h10H,2H2,1H3,(H,6,7)(H,8,9) |
| SMILES | CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 2-hydroxy-2-methylbutanedioic acid |
| Molecular Weight (g/mol) | 148.11 |
| Percent Purity | >95 |