missing translation for 'onlineSavingsMsg'
Learn More

dl-Alpha-Tocopherol, TRC

Product Code. 17992215
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992215 1 g 1g
30004959 50 g 50g
17963925 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992215 Supplier Toronto Research Chemicals Supplier No. TRCT5261451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29 H50 O2
Molecular Weight (g/mol) 430.71
Synonym 6-Chromanol, 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)- (8CI), 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-1-benzopyran-6-ol, (±)-α-Tocopherol, 2,5,7,8-Tetramethyl-2-(4',8',12'-trimethyltridecyl)chroman-6-ol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, DL-α-Tocopherol, Elementol B, Elementol Basic, Elementol R, Ephanyl, Irganox E 210, Rac-α-Tocopherol, Ronotec 201, Ronotec 202, Ronotec DF 120, Uvinul 2000AO, all-rac-α-Tocopherol, dl-α-Tocopherol
IUPAC Name 2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydrochromen-6-ol
SMILES CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3
Formula Weight 430.3811
Percent Purity >95%
Chemical Name or Material DL-alpha-Tocopherol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.