missing translation for 'onlineSavingsMsg'
Learn More

DL-5-Fluorotryptophan, MP Biomedicals

Product Code. 11452006 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452006 Supplier MP Biomedicals Supplier No. 0521017780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 154-08-5
Molecular Formula C11H11FN2O2
MDL Number MFCD00005649
Quantity 100 mg
Synonym 5-fluoro-dl-tryptophan, 5-fluorotryptophan, dl-5-fluorotryptophan, 2-amino-3-5-fluoro-1h-indol-3-yl propanoic acid, tryptophan, 5-fluoro, tryptophan, 5-fluoro-, dl, 5-fluoro-d,l-tryptophan, 2-amino-3-5-fluoroindol-3-yl propanoic acid, h-dl-trp 5-f-oh, dl-tryptophan, 5-fluoro
InChI Key INPQIVHQSQUEAJ-VIFPVBQESA-N
SMILES [NH3+][C@@H](CC1=CNC2=CC=C(F)C=C12)C([O-])=O
IUPAC Name (2S)-2-azaniumyl-3-(5-fluoro-1H-indol-3-yl)propanoate
Molecular Weight (g/mol) 222.22
PubChem CID 9577
ChEBI CHEBI:77837
Formula Weight 222.22
Chemical Name or Material Dl-5-Fluorotryptophan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.