Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
50 mL
100 mL
Material:
Duran Glass
Duran Tubing, PTFE
missing translation for 'unitSize'
Each
Description
- Receivers with straight adapter
- With cooling jacket, graduated, with socket and cone NS 29/32
- Vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Capacity (Metric) | 100 mL |
| Standard Taper Joint | Socket and Cone NS 29/32 |