Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Click to view available options
Capacity (Metric):
50 mL
100 mL
Material:
Duran Glass
Duran Tubing, PTFE
Unit Size:
Each
Description
- Receivers with straight adapter
- With cooling jacket, graduated, with socket and cone NS 29/32
- Vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Tubing, PTFE |
| Capacity (Metric) | 100 mL |
| Standard Taper Joint | Socket and Cone NS 29/32 |