Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz-Thiele, Bent
Product Code. 10030231
Click to view available options
Standard Taper Joint:
Socket and Cone NS 14/23
Socket and Cone NS 29/32
Capacity (Metric):
25 mL
50 mL
100 mL
Material:
Duran Glass
Duran PTFE
Unit Size:
Each
Product Code. 10030231
Brand: Lenz Laborglasintrumente 05298101
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Receivers with bent adapter, graduated
  • With NS socket and NS cone
  • vacuum connection: thread GL18
  • NS stopcock with solid glass or PTFE plug
  • Made of DURAN™ tubing
Specifications
Includes Bent adapter, graduated, with NS socket and NS cone, vacuum connection: thread GL18
Material Duran PTFE
Capacity (Metric) 25 mL
Standard Taper Joint Socket and Cone NS 14/23