Learn More
Lenz Laborglasintrumente Distilling Links with Splash Head
Product Code. p-7222467
Click to view available options
Length (Metric):
150 mm
200 mm
300 mm
Unit Size:
Each
Product Code. 10244341
Brand: Lenz Laborglasintrumente 05043001
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With two vertical NS cones and splash head
  • Made of DURAN™ tubing
Specifications
Length (Metric) 150 mm
Material Borosilicate Glass 3.3
Standard Taper Joint NS 14/23