Learn More
Lenz Laborglasintrumente™ Distilling Links with Liebig Condenser and GL Thread
Click to view available options
Standard Taper Joint:
Cone NS 14/23
Unit Size:
Each
Description
- Distilling links with Liebig condenser
- Outlet with NS cone
- Water connection with thread GL14
- With socket NS 14/23 for distillation thermometers of 55 mm built-in length
- Made of DURAN™ tubing
Specifications
Specifications
| Length (Metric) | 250 mm |
| Type | Distilling links with Liebig condenser |
| Material | DURAN™ tubing |
| Quantity | 1 |
| Standard Taper Joint | Cone NS 14/23 |