Learn More
Dissolved Oxygen Refilling Electrolyte for ECDOGEN-S and ECDOTPII-S DO Electrodes
Description
Eutech™ offers a wide range of Dissolved Oxygen (DO) meters and probes that measure the amount of gaseous oxygen dissolved in a solution in various educational, laboratory and industrial applications. Eutech™ Dissolved Oxygen Refilling Electrolytes are used for ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Process Electrodes.
Features:
- Refill Electrolyte Solution in 480 mL bottles
Specifications
Specifications
| Description | DO refilling electrolyte for ECDOGEN-S electrode, 480mL |
| For Use With (Equipment) | ECDOGEN-S DO Electrode |
| Type | Refilling Electrolyte Solution |
| Probe Type | DO Galvanic Electrode |