Learn More
Biochrom Disposable Visible/UV Cuvette
Click to view available options
Capacity (Metric):
70 μL
800 μL
Unit Size:
Pack of 100
Description
- Disposable cell for visible and UV spectrophotometry
- Pathlength: 10mm
Specifications
Specifications
| Type | UV Visible Cuvette |
| Disposable | Single-use |
| Capacity (Metric) | 70 μL |
| Material | UV Plastic |
| Quantity | 100 Pack |
| Path Length | 10 mm |
| For Use With (Equipment) | GeneQuant 1300 visible/UV spectrophotometer |