Learn More
Swann-Morton™ Disposable Scalpel Blade
Product Code. 12952535
Change view
Click to view available options
Quantity:
20 x 5/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12952535 20 x 5/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12952535 Supplier Swann-Morton™ Supplier No. 0113
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Non-sterile, foil-packed, disposable scalpel blade. Swann-Morton™ Disposable Scalpel Blade is constructed of carbon steel.
  • Non-sterile blade
  • Foil packets of 5
  • Fitment No.4. Fits handles 4, 4L, 4 Graduated and 6B
  • Carbon steel construction
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
For Use With (Equipment) Fitment # 4. Fits handles # 4, # 4L, # 4 Graduated and # 6B
Disposable Single-use
Sterility Non-sterile
Certifications/Compliance BS EN 7740
Quantity 20 x 5/Pk.