Learn More
Kartell™ Disposable Petri Dishes
Click to view available options
Diameter (Metric):
90 mm
Unit Size:
Pack of 500
Description
With aeration notches on the lid to prevent condensation formation
Aseptic
Polystyrene
ß Beta-ray sterilization
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Color | Clear |
| Disposable | Single-use |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Vent | No |
| Diameter (Metric) | 90 mm |