Learn More
Nexcelom™ Disposable Hemacytometer
Click to view available options
Total Count:
25 slides for 50 counts
Unit Size:
Each
Description
- An enclosed chamber with two ports for sample introduction
- The chamber contains precisely spaced lines in a grid pattern
- There are two counting chambers per slide for replicates
- The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
Specifications
Specifications
| Capacity (Metric) | 20 microliters |
| Total Count | 25 slides for 50 counts |