Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeve
Fits like a second skin without adding bulk
Brand: Lenz Laborglasinstrumente™ 01101514
Additional Details : Weight : 0.99000kg
Beschreibung
- Thin walled
- Outstanding chemical resistance at operating temperatures from -200 to +260 °C
- Without grip collar, simplifying use of joint clips
Spezifikation
50 μm | |
Disposable | |
PTFE | |
For ground joints NS 14/23 |
14/23 | |
10 | |
-200 to +260 °C |