Learn More
Poulten & Graf GmbH Discharge Tube Set for OPTIFIX™ Dispenser
Product Code. 10595692
Click to view available options
Volume (Metric):
2, 2 mL
200, 500 mL
5, 10 mL
Unit Size:
Each
Product Code. 10595692
Brand: Poulten & Graf GmbH 101 09438
Poulten & Graf GmbH Discharge Tube Set for OPTIFIX™ Dispenser is an accessory to be used with dispensers that includes a cap.
  • With stabaliser
  • Includes cap
  • Made of PTFE and PP
  • Basic and safety
Specifications
Product Type Discharge Tube for OPTIFIX Dispensers
Autoclavable Autoclavable
Description Discharge tube for OPTIFIX dispensers
Material PTFE, Polypropylene
Volume (Metric) 5, 10 mL
For Use With (Equipment) OPTIFIX dispensers
Quantity 1