Learn More
BD Discardit™ Non-Sterile Two-Part Syringe
Click to view available options
Volume (Metric):
5 mL
Graduations:
0.2 mL
Unit Size:
Pack of 3000
Description
- Two-piece syringes with Luer pip and leak-tight design
- Clear barrel with accurate graduation scale markings for perfect visualization of contents
- Ergonomically designed plunger for single-handed operation with reduced sliding force
- Silicone oil-free lubricant enables smooth, gradual plunger movement
- Retaining ring prevents accidental plunger rod withdrawal
- Design of finger grip flanges ensure stability and comfort during injection
- Single use, individually wrapped syringes are latex- and PVC-free
Specifications
Specifications
| Material | Oleamide (Lubricant), Polyethylene (Plunger), Polypropylene (Barrel) |
| Color | Clear |
| For Use With (Application) | Injection and/or aspiration of medical fluids |
| Product Type | 2-part Syringe |
| Disposable | Single-use |
| Graduations | 0.2 mL |
| Quantity | 100/Pk. |
| Volume (Metric) | 5 mL |
| Connection Type | Luer Tip |
| Sterility | Non-sterile |