Learn More
BD Discardit™ Eccentric Luer-Slip Two-Piece Syringe
Product Code. 12349199
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
20 mL
Includes:
BD Microlance Needle Packed Aside
Without Needle
Unit Size:
Pack of 100
Pack of 80
Product Code. 12349199
Brand: BD 300318
Use for sterile applications. BD Biosciences™ Discardit™ Eccentric Luer-Slip Two-Piece Syringe are disposible after single use.
  • Sterile
  • Single use
  • Two-piece syringe
  • Leur-Slip
  • Disposable
  • Clear
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene (Body)
Color Clear
Needle Gauge 21 G
Includes BD Microlance Needle Packed Aside
Product Type Eccentric Luer-slip 2-piece Syringe
Disposable Single-use
Quantity 100/Pk.
Volume (Metric) 2 mL
Connection Type Eccentric Luer Slip
Graduated Yes