missing translation for 'onlineSavingsMsg'
Learn More

DIRC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16875115 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16875115 150 μL 150µL
16885115 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16875115 Supplier Proteintech Supplier No. 267501AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Electrogenic metabolite transporter.

Specifications

Antigen DIRC2
Applications Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DIRC2
Gene Accession No. Q8BFQ6, Q96SL1
Gene Alias DIRC2, disrupted in renal carcinoma 2, RCC4, SLC49A4
Gene Symbols DIRC2
Host Species Rabbit
Immunogen DIRC2 Fusion Protein Ag24687
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224132, 84925
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.