missing translation for 'onlineSavingsMsg'
Learn More

Diphenylpyraline, TRC

Product Code. 30374290
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374290 500 mg
30380526 50 mg
30374532 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374290 Supplier Toronto Research Chemicals Supplier No. TRCD492110500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-20-6
Molecular Formula C19 H23 N O
Molecular Weight (g/mol) 281.39
Synonym Diphenylpyraline, 4-(Diphenylmethoxy)-1-methylpiperidine, 1-Methyl-4-piperidyl benzhydryl ether, 4-(Benzhydryloxy)-1-methylpiperidine, AN 1041, Diphenpyraline, Diphenylpyraline, Diphenylpyrilene, Hystryl, Lyssipoll, Mepiben, N-Methyl-4-(benzhydryloxy)piperidine, Neargal, P 253
IUPAC Name 4-benzhydryloxy-1-methylpiperidine
SMILES CN1CCC(CC1)OC(c2ccccc2)c3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H23NO/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-11,18-19H,12-15H2,1H3
Formula Weight 281.178
Percent Purity >95%
Chemical Name or Material Diphenylpyraline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.