missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfoxide, 98+%

Product Code. p-7026234
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499647 25 g 25g
11409657 100 g 100g
11419657 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499647 Supplier Thermo Scientific Alfa Aesar Supplier No. B22135.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 945-51-7 | C12H10OS | 202.271 g/mol

Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

Solubility
Soluble in water. Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.271
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 69°C to 72°C
Boiling Point 206°C to 208°C (13 mmHg)
Quantity 25 g
Beilstein 1908444
Solubility Information Soluble in water. Soluble in most organic solvents.
Formula Weight 202.28
Percent Purity ≥98%
Chemical Name or Material Diphenyl sulfoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.