missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Iodonium-2-carboxylate, MP Biomedicals

Product Code. 11481403 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481403 10 g 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11481403 Supplier MP Biomedicals Supplier No. 0522767625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 109545-72-4
Molecular Formula C13H9IO2
Quantity 10 g
Synonym diphenyliodonium-2-carboxylate, iodonium, 2-carboxyphenyl phenyl-, inner salt, 2-carboxylatophenyl phenyliodonium, 2-carboxyphenyl phenyliodonium hydroxide inner salt, iodonium, 2-carboxyphenyl phenyl-,hydroxide,innersalt, diphenyliodonium-2-carboxylate monohydrate, 98+%, 2-carboxylatophenyl phenyl iodanium, iodonium, hydroxide, inner salt, iodonium, 2-carboxyphenyl phenyl-, hydroxide, inner salt, iodonium, o-carboxyphenyl phenyl-, hydroxide, inner salt
InChI Key LUGVQQXOGHCZNN-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2C(=O)[O-]
IUPAC Name 2-phenyliodoniobenzoate
Molecular Weight (g/mol) 324.117
PubChem CID 73883
Formula Weight 342.13
Chemical Name or Material Diphenyl Iodonium-2-Carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.