Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns
BIOPHARMA
Product Code. p-4524119
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
4.6 mm
9 mm
22 mm
Particle Size:
4 μm
8 μm
Unit Size:
Each
Product Code. 11341813
Brand: Thermo Scientific™ 063000
Only null left
Add to basket
Add to basket
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
TRUSTED_SUSTAINABILITY
Specifications
Column Format Analytical Column
Diameter (Metric) 4 mm
Flow Rate 1.2 mL/min
For Use With UltiMate 3000 Titanium System
Length (Metric) 250 mm
Max. Pressure 4000 psi (275 bar)
Mobile Phase Compatibility pH 2 to 12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, perchlorate
Particle Size 8 μm
pH 2.5 to 12.5
Pore Size Nonporous
Quantity Each
Temperature 85°C
Stationary Phase Quaternary Ammonium
Product Line DNAPac
Packing Material Spherical, Nonporous Polymer
Column Type Ion Exchange
Phase Ion Exchange
Type Analytical Column
Technique Bio-LC
Green Features Responsibly packaged