Learn More
Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits
Product Code. p-4528185 Shop All Thermo Scientific Products
Click to view available options
Material:
Polystyrene
Includes:
Caps and Split Septa
Unit Size:
Pack of 100
Product Code. 11371743
Brand: Thermo Scientific™ 055058
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Use Thermo Scientific™ Dionex™ AS-AP Autosampler Vial Kits offered in polystyrene, glass or polypropylene, with caps and split septa.
Septa included are blue.
Specifications
Type Vial Kit
Includes Caps and Split Septa
Material Polystyrene
For Use With (Equipment) AS-AP Autosampler
Volume (Metric) 10 mL
Quantity Each 1