Learn More
OHAUS™ Dimpled Mat
Product Code. p-7158976 Shop All Ohaus Products
Click to view available options
Dimensions (L x W):
32.3 x 25.4 cm
35.6 x 28 cm
Unit Size:
Each
Product Code. 15722559
Brand: OHAUS™ 30400140
Only null left
Add to basket
Add to basket
OHAUS™ Dimpled Mat designed to securely hold tubes in place.
  • A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
  • Platforms can increase the amount of usable space for samples
  • In some cases can be stacked to increase throughput and double sample capacity
  • Tube racks and dimpled mats securely hold tubes in place
TRUSTED_SUSTAINABILITY
Specifications
Product Type Platform Mat
Material PVC
Depth (English) 12.75 in.
Depth (Metric) 32.40 cm
Dimensions (L x W) 32.3 x 25.4 cm
Length (English) 12.75 in.
Length (Metric) 32.40 cm
Width (English) 10.00 in.
Width (Metric) 25.40 cm
Description Dimpled Mat For Rocker SHRK04DG
For Use With (Equipment) Rocker