Learn More
Dimethoxydiphenylsilane, 97%
Product Code. 11396756
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
Product Code. 11396756
Brand: Thermo Scientific Alfa Aesar L04031.30
Request Bulk or Custom Format
CAS: 6843-66-9 | C14H16O2Si | 244.365 g/mol
Reagent for preparation of diphenylsilylene derivatives of diols by exchange.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6843-66-9
Molecular Formula C14H16O2Si
Molecular Weight (g/mol) 244.365
MDL Number MFCD00025690
InChI Key AHUXYBVKTIBBJW-UHFFFAOYSA-N
Synonym diphenyldimethoxysilane, silane, dimethoxydiphenyl, dimethoxy diphenyl silane, unii-02qb6788gc, benzene, 1,1'-dimethoxysilylene bis, diphenyldimethoxy silane, diphenyldimethoxylsilicane, diphenyl dimethoxy silane, dimethoxy-di phenyl silane, acmc-1bc5s
PubChem CID 81284
IUPAC Name dimethoxy(diphenyl)silane
SMILES CO[Si](C1=CC=CC=C1)(C2=CC=CC=C2)OC
Specifications
Boiling Point 160°C to 162°C (15mmHg)
Quantity 250 g
Beilstein 2940458
Flash Point 121°C (249°F)
Formula Weight 244.37
Percent Purity 97%
Refractive Index 1.542
Sensitivity Moisture sensitive
Density 1.079
Chemical Name or Material Dimethoxydiphenylsilane