missing translation for 'onlineSavingsMsg'
Learn More

Dihydroxy Bendamustine Sodium Salt, TRC

Product Code. 30138152
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138152 100 mg 100mg
30010546 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138152 Supplier Toronto Research Chemicals Supplier No. TRCD446565100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C16H22N3NaO4
Molecular Weight (g/mol) 343.35
Synonym 5-[Bis(2-hydroxyethyl)amino]-1-methyl-1H-benzimidazole-2-butanoic Acid Sodium Salt;
SMILES OCCN(CCO)C1=CC=C(N(C)C(CCCC(O[Na])=O)=N2)C2=C1

Specifications

Analyte or Component Names Dihydroxy Bendamustine Sodium Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C16H23N3O4/c1-18-14-6-5-12(19(7-9-20)8-10-21)11-13(14)17-15(18)3-2-4-16(22)23/h5-6,11,20-21H,2-4,7-10H2,1H3,(H,22,23)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Chemical Name or Material Dihydroxy Bendamustine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.